US
EN
Applications
Products
Services
Support
Home
Search Results
(±)−α−Tocopherol (Vitamin E) solution
(1)
(±)−α−Tocopherol (Vitamin E) solution
Synonyms:
(±)-α-Tocopherol, Vitamin E, DL-all-
rac
-α-Tocopherol
Empirical Formula (Hill Notation):
C
29
H
50
O
2
CAS Number:
10191-41-0
Molecular Weight:
430.71
EC Number:
200-659-6
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
V-020
(±)−α−Tocopherol (Vitamin E) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant
®
Expand
View Pricing