US
EN
Applications
Products
Services
Support
Home
Search Results
Amphotericin B solution
(1)
Amphotericin B solution
Empirical Formula (Hill Notation):
C
47
H
73
NO
17
CAS Number:
1397-89-3
Molecular Weight:
924.08
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
A2942
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Expand
View Pricing