US
EN
Applications
Products
Services
Support
Home
Search Results
(±)-Amphetamine-d
6
solution
(2)
(±)-Amphetamine-d
6
solution
Empirical Formula (Hill Notation):
C
9
D
6
H
7
N
CAS Number:
73758-26-6
Molecular Weight:
141.24
EC Number:
200-659-6
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
A-045
(±)-Amphetamine-d
6
solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant
®
Expand
View Pricing
A-044
(±)-Amphetamine-d
6
solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant
®
Expand
View Pricing