MilliporeSigma
(±)-Amphetamine-d5 (deuterium label on side chain) solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

(±)-Amphetamine-d5 (deuterium label on side chain) solution

(±)-Amphetamine-d5 (deuterium label on side chain) solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C9D5H8N
CAS Number:
Molecular Weight:
140.24

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
(±)-Amphetamine-d5 (deuterium label on side chain) solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
(±)-Amphetamine-d5 (deuterium label on side chain) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®