US
EN
Applications
Products
Services
Support
Home
Search Results
Amberlite
®
IR120 Na+ form
(1)
Amberlite
®
IR120 Na+ form
CAS Number:
68441-33-8
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
224359
Amberlite
®
IR120 Na+ form, Na
+
-form
Expand
View Pricing