MilliporeSigma
(±)-11-nor-9-Carboxy-Δ9-THC solution 100 μg/mL in methanol, ampule of 1 mL, Cerilliant®

(±)-11-nor-9-Carboxy-Δ9-THC solution

(±)-11-nor-9-Carboxy-Δ9-THC solution 100 μg/mL in methanol, ampule of 1 mL, Cerilliant®
Empirical Formula (Hill Notation):
C21H28O4
CAS Number:
Molecular Weight:
344.44
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
(±)-11-nor-9-Carboxy-Δ9-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
(±)-11-nor-9-Carboxy-Δ9-THC solution, 100 μg/mL in methanol, ampule of 1 mL, Cerilliant®