Products
US
EN
Products
Applications
Products
Services
Support
Home
Search Results
(±)-11-nor-9-Carboxy-Δ
9
-THC-D
3
solution
(2)
(±)-11-nor-9-Carboxy-Δ
9
-THC-D
3
solution
Empirical Formula (Hill Notation):
C
21
H
25
O
4
D
3
CAS Number:
136844-96-7
Molecular Weight:
347.46
EC Number:
200-659-6
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
T-008
(±)-11-nor-9-Carboxy-Δ
9
-THC-D
3
solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant
®
Expand
View Pricing
T-004
(±)-11-nor-9-Carboxy-Δ
9
-THC-D
3
solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant
®
Expand
View Pricing