Products
US
EN
Products
Applications
Products
Services
Support
Home
Search Results
(±)-11-Hydroxy-Δ
9
-THC solution
(2)
(±)-11-Hydroxy-Δ
9
-THC solution
Empirical Formula (Hill Notation):
C
21
H
30
O
3
CAS Number:
34675-49-5
Molecular Weight:
330.46
EC Number:
200-659-6
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
H-027
(±)-11-Hydroxy-Δ
9
-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant
®
Expand
View Pricing
H-026
(±)-11-Hydroxy-Δ
9
-THC solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant
®
Expand
View Pricing