Product Name
RPMI-1640 Medium With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Product Number
R0883
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH7.0 - 7.6
Osmolality264 - 292 mOs/kg
Cell Culture Testing - MTTPass
Cell LineCell Line - Cell Types
SterilityPass
Endotoxin Level<= 1 EU/ml
Glucose Concentration1.8 - 2.2 g/l
Cloning AssayPass