Product Name
Minimum Essential Medium Eagle With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Product Number
M4655
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH7.0 - 7.6
Osmolality286 - 317 mOs/kg
Cell Culture Testing - MTTPass
Cell LineCell Line - Cell Types
SterilityPass
Endotoxin Level<= 1 EU/ml
Glucose Concentration0.9 - 1.1 g/l