Product Name
Dulbecco’s Modified Eagle’s Medium - low glucose With 1000 mg/L glucose, L-glutamine, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Product Number
D6046
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH7.0 - 7.6
Osmolality308 - 340 mOs/kg
Cell Culture Testing - MTTPass
Cell LineCell Line - Cell Types
SterilityPass
Endotoxin Level<= 1 EU/ml
Glucose Concentration0.9 - 1.1 g/l