Product Name
Dulbecco’s Modified Eagle’s Medium - high glucose With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Product Number
D5671
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH7.0 - 7.6
Osmolality317 - 351 mOs/kg
Cell Culture Testing - MTTPass
Cell LineCell Line - Cell Types
Cloning AssayPass
SterilityPass
Endotoxin Level<= 1 EU/ml
Glucose Concentration4.05 - 4.95 g/l
Animal Derived Component FreeConforms