Product Name
Hexamethyldisilazane produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Product Number
86944
Product Brand
SIAL
CAS Number
999-97-3
Molecular Weight
161.39
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 97.0 %
REFRACTIVE INDEX N20/D1.407 - 1.409
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE