MilliporeSigma
All Photos(1)

K08670383

Kromasil® Silica, Bulk

10 μm particle size, pore size 60 Å, pkg of 500 g

EC Number:
NACRES:
SB.52

product line

Kromasil®

form

powder

packaging

pkg of 500 g

manufacturer/tradename

Kromasil® KR60-10-SIL-500GM

availability

available only in USA, Canada and Puerto Rico

matrix active group

silica phase

particle size

10 μm

pore size

60 Å pore size

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

Legal Information

Kromasil is a registered trademark of EKA Chemicals AB

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service