All Photos(1)

58281

Supelco

Normal Phase Test Mix

in methylene chloride (varied conc.), analytical standard

EC Number:

grade

analytical standard

Quality Level

packaging

ampule of 1 mL

concentration

in methylene chloride (varied conc.)

application(s)

HPLC: suitable
gas chromatography (GC): suitable

compatibility

use to QA LC-Si (silica) columns

format

multi-component solution

storage temp.

room temp

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Analyte

Description

    Acetanilide 20 μg/mL
    Benzanilide 20 μg/mL
    Benzene 600 μg/mL

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service