MilliporeSigma
All Photos(1)

48182

Supelco

ASTM® D5307 Crude oil qualitative standard

analytical standard

grade

analytical standard

Quality Level

Agency

ASTM® D5307

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

(each component in approx proportion indicated)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

General description

ASTM® D5307 Crude oil qualitative standard is an analytical standard used for determining the boiling range distribution of crude petroleum.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Each of the hydrocarbons listed in approximate proportions (w/w) indicated. Combine with Cat. No. 48179 to prepare a calibration mixture covering C3 to C44 for calibrating retention times for boiling point correlation.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Butane 15 % (w/w)
    Heptane 15 % (w/w)
    Hexane 15 % (w/w)
    Nonane 15 % (w/w)
    Octane 15 % (w/w)
    Pentane 15 % (w/w)
    Propane 10 % (w/w)

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-5.8 °F - closed cup

Flash Point(C)

-21 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service