MilliporeSigma
All Photos(1)

48157

Supelco

Indole solution

2000 μg/mL in methanol, certified reference material

CAS Number:

Quality Level

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C8H7N/c1-2-4-8-7(3-1)5-6-9-8/h1-6,9H

InChI key

SIKJAQJRHWYJAI-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Stephen J Heffernan et al.
Chemical communications (Cambridge, England), 49(23), 2314-2316 (2013-02-13)
The Au(I)-catalysed rearrangement of propargylic esters formed from an ynamide has been studied. The reaction is facile, and when conducted in the presence of a reactive indole nucleophile, leads to a cascade process whereby γ-indolyl α-acyloxyenamides are formed in good
Vida Mashayekhi et al.
Chemical & pharmaceutical bulletin, 61(2), 144-150 (2012-11-17)
Based on the existing reports regarding the antiplatelet aggregation activity of hydrazone derivatives, a series of indole hydrazone derivatives were considered as potential antiplatelet agents and synthesized. The structures of the synthesized compounds were confirmed by spectral data and elemental
Israel Alvarado et al.
Journal of bacteriology, 195(5), 1005-1011 (2012-12-25)
Paenibacillus larvae is the causative agent of American foulbrood (AFB), a disease affecting honey bee larvae. First- and second-instar larvae become infected when they ingest food contaminated with P. larvae spores. The spores then germinate into vegetative cells that proliferate
Facile access to 3-acylindoles through palladium-catalyzed addition of indoles to nitriles: the one-pot synthesis of indenoindolones.
Yuanhong Ma et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(4), 1189-1193 (2012-12-15)
Kap-Sun Yeung et al.
Bioorganic & medicinal chemistry letters, 23(1), 198-202 (2012-12-04)
A series of substituted carboxamides at the indole C7 position of the previously described 4-fluoro-substituted indole HIV-1 attachment inhibitor 1 was synthesized and the SAR delineated. Heteroaryl carboxamide inhibitors that exhibited pM potency in the primary cell-based assay against a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service