MilliporeSigma
All Photos(1)

47559-U

Supelco

Linseed (flaxseed) oil

analytical standard

Synonym(s):
Linseed oil
CAS Number:
EC Number:
MDL number:
NACRES:
NA.24

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.4795 (lit.)

density

0.93 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Characterized linseed (flaxseed) oil samples are available for use as controls or check samples for fatty acid methyl ester (FAME) analyses. These reference materials can be used as qualitative standards. They provide an excellent means of standardizing the lipid procedures and comparing the results to others. Packed in an amber ampule under nitrogen. A Certificate of Composition, which includes the chromatographic fingerprint analysis, is provided with the oil sample.
This reference material is offered as a qualitative standard. Use the chromatographic fingerprint of our reference material to check for the presence of linseed oil in your sample. A Certificate of Composition which includes a chromatogram is shipped with each product.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service