MilliporeSigma
All Photos(1)

41979-U

Supelco

EPA TO-15/17 Calibration mix

25 components, 100 ppb each component in nitrogen, cylinder of 110 L (pi-marked cylinder), analytical standard

EC Number:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA TO-15,TO-17

feature

standard type calibration

packaging

cylinder of 110 L (pi-marked cylinder)

manufacturer/tradename

Air Liquide America Specialty Gases LLC 0104AP00002 110

concentration

100 ppb each component in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Related Categories

General description

EPA TO-15/17 Calibration mix [25 components, 100 ppb each component in nitrogen, cylinder of 110 L (pi-marked cylinder)] may be used as a chemical reference standard for the qualitative calibration of air monitoring and industrial hygiene equipment. These standards are available in both liquid solutions and gaseous blends, to accommodate calibration of today′s variety of air monitoring and analysis devices and applications. Each standard includes a Certificate of Analysis summarizing the testing data. Each gaseous standard is provided in a transportable cylinder for ease of use in either the laboratory or the field.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

The Scotty TO-15/17 gas calibration mix is supplied in an Extra-Life transportable aluminum cylinder which features Air Liquide′s exclusive ACULIFE cylinder treatment process for guaranteed mixture stability.

Blend tolerance and analytical accuracy = +/- 10% for all components except benzyl chloride, carbon disulfide, 2-propanol which are +/- 25%.

Specifications

Cylinder specifications:
Pressure:1800 psig
Outlet fitting:CGA 180
Weight:2.2lbs. (1kg)
Dimensions:3.25in D X 11.625in H (82.6mm X 295.3mm)

U.S. D.O.T. specs: 3AL2216

Legal Information

ACULIFE is a trademark of Air Liquide

Analyte

Description

    Acetone
    Allyl chloride
    Benzyl chloride
    Bromodichloromethane
    Bromoform
    1,3-Butadiene
    2-Butanone
    Carbon disulfide
    Cyclohexane
    Dibromochloromethane
    1,4-Dioxane
    Ethyl acetate
    4-Ethyltoluene
    Heptane
    Hexane
    2-Hexanone
    4-Methyl-2-pentanone
    tert-Butyl methyl ether
    2-Propanol
    Propylene
    Tetrahydrofuran
    trans-1,2-Dichloroethylene
    2,2,4-Trimethylpentane
    Vinyl acetate
    Vinyl bromide
    See All (25)

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service