MilliporeSigma
All Photos(1)

41975-U

Supelco

Ozone Precursor / PAMS Mix

57 components, 100 ppb each component in nitrogen, cylinder of 110 L (pi-marked cylinder), analytical standard

EC Number:
NACRES:
NA.24

grade

analytical standard

Quality Level

feature

standard type calibration

packaging

cylinder of 110 L (pi-marked cylinder)

manufacturer/tradename

Air Liquide America Specialty Gases LLC TAZ00005-110

concentration

100 ppb each component in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Related Categories

General description

Ozone Precursor / PAMS Mix [57 components, 100 ppb each component in nitrogen, cylinder of 110 L (pi-marked cylinder)] manufactured by Air Liquide America Speciality Gases Toxic Organic (TO) is an excellent choice of calibration gas mixtures for use in monitoring ppb levels of toxic volatiles and semi-volatiles in air in accordance with U.S. Environmental Protection Agency′s TO-15/17 and Photochemical Assessment Monitoring Systems (PAMS) methodologies, and other related government regulations. The gas mixture is produced individually and gravimetrically using NIST traceable weights.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

The Scotty Ozone Precursor/PAMS gas calibration mix is supplied in an Extra-Life transportable aluminum cylinder which features Air Liquide′s exclusive ACULIFE cylinder treatment process for guaranteed mixture stability.

Blend tolerance and analytical accuracy = +/- 10%.

Specifications

Cylinder specifications:
Pressure:1800 psig
Outlet fitting:CGA 180
Weight:2.2lbs. (1kg)
Dimensions:3.25in D X 11.625in H (82.6mm X 295.3mm)

U.S. D.O.T. specs: 3AL2216

Legal Information

ACULIFE is a trademark of Air Liquide

Analyte

Description

    Acetylene
    Benzene
    Butane
    1-Butene
    cis-2-Butene
    trans-2-Butene
    Cumene
    Cyclohexane
    Cyclopentane
    Decane
    1,3-Diethylbenzene
    1,4-Diethylbenzene
    2,2-Dimethylbutane
    2,3-Dimethylbutane
    2,3-Dimethylpentane
    2,4-Dimethylpentane
    Dodecane
    Ethane
    Ethylbenzene
    Ethylene
    3-Ethyltoluene
    2-Ethyltoluene
    4-Ethyltoluene
    Heptane
    Hexane
    1-Hexene
    Isoprene
    Mesitylene
    2-Methylpropane
    2-Methylbutane
    Methylcyclohexane
    Methylcyclopentane
    2-Methylheptane
    3-Methylheptane
    2-Methylhexane
    3-Methylhexane
    2-Methylpentane
    3-Methylpentane
    Nonane
    Octane
    Pentane
    1-Pentene
    cis-2-Pentene
    trans-2-Pentene
    Propane
    Propylbenzene
    Propylene
    Styrene
    Toluene
    1,2,3-Trimethylbenzene
    1,2,4-Trimethylbenzene
    2,2,4-Trimethylpentane
    2,3,4-Trimethylpentane
    Undecane
    o-Xylene
    m/p-xylene
    See All (56)

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service