MilliporeSigma
All Photos(1)

41973-U

Supelco

EPA TO-15/17 Calibration mix

62 components, 1 ppm in nitrogen, cylinder of 110 L (pi-marked cylinder), analytical standard

EC Number:
NACRES:
NA.24

grade

analytical standard

Agency

EPA TO-15,TO-17

shelf life

12 mo.

feature

standard type calibration

packaging

cylinder of 110 L (pi-marked cylinder)

manufacturer/tradename

Air Liquide America Specialty Gases LLC TAZ70002-110

concentration

1 ppm in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

General description

EPA TO-15/17 Calibration mix is a quantitative formulation for use as a chromatographic calibration standard for the determination of volatile organic compounds (VOCs) by gas chromatography-mass spectrometry (GC-MS). It may also be used as a chemical reference standard for the qualitative calibration of air monitoring and industrial hygiene equipment.
These standards are available in both liquid solutions and gaseous blends, to accommodate calibration of today′s variety of air monitoring and analysis devices and applications. Each standard includes a Certificate of Analysis summarizing the testing data. Each standard solution is provided in a flame-sealed amber ampoule. Each gaseous standard is provided in a transportable cylinder for ease of use in either the laboratory or the field.

Application

EPA TO-15/17 Calibration mix (62 components, 1 ppm in nitrogen) may be used as an analytical standard to quantify volatile organic compounds (VOCs) in food waste emissions as well as ambient and atmospheric air samples by GC-MS.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

The Scotty TO-15/17 gas calibration mix is supplied in an Extra-Life transportable aluminum cylinder which features the exclusive ACULIFE cylinder treatment process for guaranteed mixture stability.

Blend tolerance and analytical accuracy = +/- 10% for all components except benzyl chloride, carbon disulfide, ethanol, 2-propanol which are +/- 25%.

Specifications

Cylinder specifications:
Pressure:1800 psig
Outlet fitting:CGA 180
Weight:2.2lbs. (1kg)
Dimensions:3.25in D X 11.625in H (82.6mm X 295.3mm)

U.S. D.O.T. specs: 3AL2216

Legal Information

ACULIFE is a trademark of Air Liquide

Analyte

Description

    Acetone
    Benzene
    Benzyl chloride
    Bromoform
    Bromomethane
    Bromodichloromethane
    1,3-Butadiene
    2-Butanone
    Carbon disulfide
    Carbon tetrachloride
    Chlorobenzene
    Chloroethane
    Chloroform
    Cyclohexane
    Chloromethane
    Dibromochloromethane
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    1,1-Dichloroethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    trans-1,2-Dichloroethylene
    1,2-Dichloropropane
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    1,4-Dioxane
    Ethanol
    Ethyl acetate
    Ethylbenzene
    1,2-Dibromoethane
    4-Ethyltoluene
    Trichlorofluoromethane
    Dichlorodifluoromethane
    1,1,2-Trichloro-1,2,2-trifluoroethane
    1,2-Dichlorotetrafluoroethane
    Heptane
    Hexachloro-1,3-butadiene
    Hexane
    2-Hexanone
    4-Methyl-2-pentanone
    Dichloromethane
    tert-Butyl methyl ether
    2-Propanol
    Propylene
    Styrene
    1,1,2,2-Tetrachloroethane
    Tetrachloroethylene
    Tetrahydrofuran
    Toluene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    1,2,4-Trichlorobenzene
    1,2,4-Trimethylbenzene
    Mesitylene
    Vinyl acetate
    Vinyl chloride
    m-Xylene
    o-Xylene
    p-Xylene
    See All (62)

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Spatiotemporal Variations and Health Implications of Hazardous Air Pollutants in Ulsan, a Multi-Industrial City in Korea
Baek KM, et al.
Chemosphere, 11(5), 547-547 (2020)
Reduction of ammonia and volatile organic compounds from food waste-composting facilities using a novel anti-clogging biofilter system
Ryu HW, et al.
Bioresource Technology, 102(7), 4654-4660 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service