All Photos(1)

41901

Supelco

EPA TO-14A Aromatic Subset Mix

100 ppb in nitrogen, pkg of 110 L (pi-marked cylinder), analytical standard

EC Number:

Quality Level

grade

analytical standard

Agency/Method

EPA TO-14 (volatile organic compounds)

usage

sufficient for ≥80 uses

packaging

pkg of 110 L (pi-marked cylinder)

concentration

100 ppb in nitrogen

parameter

1800 psi pressure

application(s)

HPLC: suitable
gas chromatography (GC): suitable

cylinder H × diam.

12 1/4  in. (31 cm) × 3 1/4  in. (8.3 cm)

fitting

CGA 180 fitting

Featured Industry

Environmental

format

multi-component solution

Related Categories

General description

EPA TO-14A Aromatic Subset Mix [100 ppb in nitrogen, pkg of 110 L (pi-marked cylinder)] may be used as a chemical reference standard for the qualitative calibration of air monitoring and industrial hygiene equipment. These standards are available in both liquid solutions and gaseous blends, to accommodate calibration of today′s variety of air monitoring and analysis devices and applications. Each standard includes a Certificate of Analysis summarizing the testing data. Each standard solution is provided in a flame-sealed amber ampoule. Each gaseous standard is provided in a transportable cylinder for ease of use in either the laboratory or the field.
These high pressure, transportable gas calibration blends for US EPA Method TO-14 ensure that laboratories can perform Method TO-14 and Clean Air Act analyses at a fraction of their former costs. Scott TO-14 gas blends are packaged in SCOTTY HP high pressure, transportable, laboratory-size cylinders. Scott TO-14 gas calibration blends are traceable to NIST reference mixtures (NIST SRM 1804) and are certified by Scott Gases for superior stability and accuracy.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

14 analytes, 100ppb each in nitrogen

Analyte

Description

    Benzene
    Chlorobenzene
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    Ethylbenzene
    Styrene
    Toluene
    1,2,4-Trichlorobenzene
    1,2,4-Trimethylbenzene
    Mesitylene
    o-Xylene
    m-Xylene
    p-Xylene
    See All (14)

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service