MilliporeSigma
All Photos(1)

29756-U

Supelco

Carbopack X

O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea

Synonym(s):
Carbopack X
NACRES:
NB.24

material

glass TD tube

Quality Level

Agency

EPA TO-17

product line

Carbopack

feature

fritted
preconditioned

packaging

pkg of 10 ea

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Carbopack X graphitized carbon black (GCB)

suitability

suitable for (for sampling 1,3-butadiene)

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for analyte group C3-C5
for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
mode of use adsorbent

Looking for similar products? Visit Product Comparison Guide

Legal Information

Carbopack is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service