27323

Supelco

Fisons 2.5mL Vial, 12 X 40mm

clear glass vial, closure type, crimp top vial, pkg of 1000 ea

Synonym(s):
Fisons Autosampler Vials

material

clear glass vial

feature

closure type crimp top vial

packaging

pkg of 1000 ea

O.D. × H

12 mm × 40 mm

volume

2.5 mL

compatibility

for use with Amersham LKB 2157-020
for use with Fisons AS 800
for use with ICI Instruments LC 1600

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

These vials are designed for use in Fisons Instruments AS 800, ICI Instruments LC1600, and Pharmacia LKB 2157-020.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service