MilliporeSigma
All Photos(1)

SAB4700390

Sigma-Aldrich

Monoclonal Anti-CD11c antibody produced in mouse

clone BU15, purified immunoglobulin, buffered aqueous solution

Synonym(s):
Anti-ITGAX
NACRES:
NA.41

Quality Level

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BU15, monoclonal

form

buffered aqueous solution

species reactivity

canine, monkey, human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... ITGAX(3687)

General description

The antibody BU15 reacts with CD11c (alphaX, p150), a 150 kDa integrin expressed mainly on dendritic cells and tissue macrophages.

Immunogen

Dendritic cells of synovial fluid

Application

The reagent is designed for Flow Cytometry analysis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Christopher Lupfer et al.
Journal of virology, 88(16), 8946-8955 (2014-05-30)
Nucleotide oligomerization and binding domain (NOD)-like receptors (NLRs) are important in the innate immune response to viral infection. Recent findings have implicated NLRP3, NOD2, and NLRX1 as important players in the innate antiviral response, but their roles in the generation
Yasuko Aida et al.
International journal of biological sciences, 10(8), 882-894 (2014-08-30)
Alveolar macrophages (AMs) play important roles in the pathogenesis of chronic obstructive pulmonary disease (COPD). We previously demonstrated upregulation of the transcription factor MafB in AMs of mice exposed to cigarette smoke. The aim of this study was to elucidate
David G Hancock et al.
PloS one, 9(6), e100613-e100613 (2014-06-21)
Dendritic cells (DCs) are critical for regulating CD4 and CD8 T cell immunity, controlling Th1, Th2, and Th17 commitment, generating inducible Tregs, and mediating tolerance. It is believed that distinct DC subsets have evolved to control these different immune outcomes.
Moonkyu Lee et al.
Archives of pharmacal research, 37(9), 1234-1240 (2014-04-22)
Myeloid-derived suppressor cells (MDSCs) accumulate in cancer patients and tumor-bearing mice, subsequently suppressing the host immune system. MDSCs represent a group of immature myeloid cells expressing CD11b and Gr-1. Here, we show that a Toll-like receptor (TLR) agonist, resiquimod, which
Chanchal Sadhu et al.
Journal of immunoassay & immunochemistry, 29(1), 42-57 (2007-12-18)
Functions and binding properties of four CD11c-specific mAbs are described here. The mAb 496B stimulated, while 496K inhibited ligand binding of CD11c. The stimulatory mAb, 496B, as well as the inhibitory mAbs BU15 and 496 K appear to act allosterically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service