All Photos(1)

745C2

87030901

biological source

mouse blood

growth mode

Suspension

karyotype

2n = 22

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse erythroleukaemic lymphoblast

Cell Line Description

This cell line is a clone isolated from the parent line 745 TG-12. The cells are resistant to oubain and thioguanine and deficient in HGPRT. Haemoglobulin synthesis can be induced with 1.5% DMSO or 1mM oubain.

Application

Haematopoiesis studies

Culture Medium

Ham′s F10 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service