All Photos(1)

C32 (colon)

12022908

Synonym(s):
C 32, C-32

biological source

human colon

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

receptors

Not specified

Looking for similar products? Visit Product Comparison Guide

Related Categories

Cell Line Origin

Human colorectal adenocarcinoma, well differentiated, Dukes′ stage C

Cell Line Description

The C32 cell line was established from a 66-year old female patient with a well differentiated adenocarcinoma of the colon classified as Dukes′ stage C.

DNA Profile

STR-PCR Data:

Amelogenin: X
CSF1PO: 10,11
D13S317: 8,11
D16S539: 9
D5S818: 11
D7S820: 9
THO1: 8,9
TPOX: 8,9
vWA: 14,18

Culture Medium

Iscove′s Modified Dulbecco′s Medium, + 10% FBS + 2 mM Glutamine

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37 °C.

C32 cells grow very slowly (see growth curve); following resuscitation or subculture the cells take at least 48 hours to re-attach. Cells should be left without disturbance during this time to facilitate adhesion. Centrifugation of the cells (100g x 5 min) at resuscitation to remove DMSO improves the establishment of a viable culture. Once attached, the cells grow in discrete islands and use of trypsin or trypsin/EDTA to subculture the cells (even without knocking the flask) yields large clumps. Further disaggregation may be achieved by repeatedly pipetting the cells.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service