All Photos(1)

P9203

Sigma-Aldrich

Polyoxyethylenesorbitan Monolaurate

suitable for electrophoresis, 20% in H2O, suitable for Western Blotting and biotinylation of proteins

Synonym(s):
TWEEN 20, Tween 20, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate
CAS Number:
MDL number:

mol wt

~1228

concentration

20% in H2O

technique(s)

electrophoresis: suitable

refractive index

n20/D 1.468 (lit.)

density

1.095 g/mL at 25 °C (lit.)

suitability

suitable for Western Blotting and biotinylation of proteins

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-ionic detergent

Legal Information

TWEEN is a registered trademark of Croda International PLC

Certificate of Analysis

Certificate of Origin

Properties of detergents.
A Helenius et al.
Methods in enzymology, 56, 734-749 (1979-01-01)
Julie Beegle et al.
Stem cells (Dayton, Ohio), 33(6), 1818-1828 (2015-02-24)
Mesenchymal stem cells/multipotent stromal cells (MSCs) are promising therapeutics for a variety of conditions. However, after transplantation, cell retention remains extremely challenging. Given that many hypoxic signals are transitory and that the therapeutic administration of MSCs is typically into tissues
Daniel C Zielinski et al.
Nature communications, 6, 7101-7101 (2015-06-10)
Drug side effects cause a significant clinical and economic burden. However, mechanisms of drug action underlying side effect pathogenesis remain largely unknown. Here, we integrate pharmacogenomic and clinical data with a human metabolic network and find that non-pharmacokinetic metabolic pathways

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service