MilliporeSigma
All Photos(1)

59429C

SAFC

Trypsin (1X)

sterile; γ-irradiated, 0.25% porcine trypsin (1:250) in HBSS, w/0.1% EDTA-Na2, w/o Ca++ and Mg++

Synonym(s):
Trypsin-EDTA solution
MDL number:
NACRES:
NA.75

Quality Level

description

for research or for further manufacturing use

sterility

sterile; γ-irradiated

form

liquid

technique(s)

cell culture | mammalian: suitable

components

phenol red: 10.620 mg/L
NaHCO3: 350.000 mg/L

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Trypsin-EDTA solutions are used to release adherent cells from culture surfaces. They are generally composed of variable concentrations of natural porcine pancreas-derived trypsin and EDTA. Trypsin 1X solutions can range from 0.025% to 0.5%. The reasons for the range of concentrations are differences in required trypsin activity or potency; differences in incubation times; and cell line susceptibility. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture. Various formulations should be tested to determine the best product for a specific application.

Preparation Note

SAFC Biosciences has validated a process using γ radiation to significantly reduce the risks associated with adventitious agents such as Porcine Parvovirus (PPV), PorcineRespiratory and Reproductive Syndrome (PRRS) and Mycoplasma hyorhinis while maintaining product performance. The use of γ irradiated trypsin requires no change to the end user′s methods or procedures while giving additional assurance against microbial contaminants associated with animal-derived products. All trypsin is obtained from the United States or other countries deemed free of Bovine Spongiform Encephalopathy (BSE).
This product does contain phenol red. Due to shipment on dry ice, there could be significant carbon dioxide buildup in the package. This CO2 may enter the solution and lower the pH slightly, giving an orange rather than pinkish color. The orange solution will still be suitable for use, or the pH can be adjusted with sodium hydroxide. Incubating cells with too high a trypsin concentration for a long period can damage cell membranes and kill the cells. Solubilizing trypsin or diluting it from a concentrated solution should be done with a buffered salt solution containing no Ca2+ or Mg2+.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service