All Photos(1)

51923

Sigma-Aldrich

FITC-Dextran sulfate sodium salt

average mol wt 500,000

NACRES:
NA.32

Quality Level

mol wt

average mol wt 500,000

composition

sulfur content, 16-20%

extent of labeling

0.001-0.020 mol/mol FITC glucose

loss

≤10% loss on drying

suitability

conforms to structure for Infrared spectrum

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Chao Niu et al.
Autophagy, 15(5), 843-870 (2019-01-18)
Studies regarding macroautophagic/autophagic regulation in endothelial cells (ECs) under diabetic conditions are very limited. Clinical evidence establishes an endothelial protective effect of metformin, but the underlying mechanisms remain unclear. We aimed to investigate whether metformin exerts its protective role against
Yun Xia et al.
Cell, 175(4), 1059-1073 (2018-10-03)
Motivated by the clinical observation that interruption of the mevalonate pathway stimulates immune responses, we hypothesized that this pathway may function as a druggable target for vaccine adjuvant discovery. We found that lipophilic statin drugs and rationally designed bisphosphonates that

Articles

FITC-labelled polysaccharides

Additional information about FITC-labelled polysaccarides including FITC-Dextran, FITC-DEAE-dextran (FDD), FITC-carboxymethyl-dextran (FCM-Dextran). This page discusses the impact of weight-average molecular weight on the respective molecule dimensions and other physical properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service