MilliporeSigma
All Photos(1)

PE1250

Supelco

Volatile Organic Compounds 1 - WP

Proficiency Testing Material, Concentrate for dilution to 100 mL

Synonym(s):
WP - VOCs
NACRES:
NA.24

Quality Level

packaging

ampule of 2 × 1.2 mL

concentration

in methanol

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

Volatile Organic Compounds 1 - WP proficiency testing material is produced in accordance with ISO/IEC 17043:2010. Also referred to as non-Potable water or Water Pollution sample, this sample is produced according to TNI requirements. However, please note, this sample may include non-TNI analytes as well. The higher criteria from the TNI tables mimic the levels common to wastewater samples. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Most water PT samples are available as concentrates or ready-to-use volumes giving the end-user flexibility in selecting samples that will fit their exact needs. Our WP program is designed to meet the quality assurance requirements, thereby ensuring the analytical competence of your laboratory.
These samples are typically provided in duplicate for use in case of lab mishap, for QC purposes or to analyze multiple methods.

Type of PT Scheme: Water analysis - Water Pollution (WP)

Matrix: Water Pollution/Wastewater

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of Volatile Organic Compounds 1 - WP can be found in the reporting packet which will be available to download at PT Portal.

Application

Volatile Organic Compounds 1 - WP is a non-Potable water (NPW) PT standard, suitable for volatile organic compound testing in water pollution/wastewater.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Features and Benefits

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Preparation Note

Description

  • This PT sample is packaged as a 1.2 mL concentrate.
  • The solvent for this sample is methanol.
  • The PT sample is not preserved.

Sample Preparation

  • Fill a 100 mL Class A Volumetric Flask to volume with organic-free reagent water.
  • Open ampule PE1250-1.2ML and withdraw 20 μL of the solution using a gas-tight microsyringe.
  • Inject the concentrate into the expanded area of the flask well below the surface of the water.
  • Stopper the flask and mix by gentle inversion 3 times. Excessive mixing will result in loss of volatiles and should be avoided.
  • This represents the sample for analysis.
  • DO NOT USE a matrix simulator with this sample.

Storage and Stability

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released unti the testing period concludes.

Proficiency Testing Schedule (2021-2022)

Wastewater
WP21-2March 24 - May 7, 2021
WP21-4September 22 - November 5, 2021

WP22-2March 23 - May 6, 2022
WP22-4September 21 - November 4, 2022

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    1,2,4-Trimethylbenzene 10-120 μg/L
    1,2-Dichlorobenzene 10-120 μg/L
    1,3,5-Trimethylbenzene 10-120 μg/L
    1,3-Dichlorobenzene 10-120 μg/L
    1,4-Dichlorobenzene 10-120 μg/L
    Benzene 10-120 μg/L
    Ethylbenzene 10-120 μg/L
    m+p-Xylene 10-150 μg/L
    Methyl tert-butyl ether (MTBE) 15-150 μg/L
    Naphthalene 15-150 μg/L
    o-Xylene 10-150 μg/L
    Toluene 10-120 μg/L
    Xylene, Total 20-300 μg/L
    See All (13)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup - Solvent

Flash Point(C)

11 °C - closed cup - Solvent

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service