MilliporeSigma
All Photos(1)

CRM023

Supelco

Trace Metals - Sandy Loam 7

certified reference material, pkg of 50 g

NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM023-050

technique(s)

AAS: suitable
ICP: suitable

pH

7.40

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

The following elements are not certified and are included for information only:
  • B, Si
Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Aluminum
    Arsenic
    Barium
    Beryllium
    Boron
    Cadmium
    Calcium
    Chromium
    Cobalt
    Copper
    Iron
    Lead
    Magnesium
    Manganese
    Mercury
    Nickel
    Potassium
    Selenium
    Silicon
    Sodium
    Strontium
    Thallium
    Vanadium
    See All (23)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service