MilliporeSigma
All Photos(1)

ARK2161

SAFC

Sodium butyrate

Synonym(s):
Butyric acid sodium salt
Linear Formula:
CH3CH2CH2COONa
CAS Number:
Molecular Weight:
110.09
Beilstein:
3629439
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder

mp

250-253 °C (lit.)

suitability

suitable for manufacturing use

storage temp.

room temp

SMILES string

[Na+].CCCC([O-])=O

InChI

1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1

InChI key

MFBOGIVSZKQAPD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service