MilliporeSigma
All Photos(1)

82524

Millipore

Protein Hydrolysate N-Z-Amine® AS

suitable for microbiology

Synonym(s):
Peptone from casein
CAS Number:
EC Number:
NACRES:
NA.85

Quality Level

biological source

bovine milk

assay

≥12% total nitrogen (N) basis
≥5% amino-nitrogen basis

form

powder

ign. residue

≤10%

loss

≤8% loss on drying

pH

6.9±0.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, faintly yellow to brown

application(s)

food and beverages
microbiology

Looking for similar products? Visit Product Comparison Guide

Legal Information

N-Z-Amine is a registered trademark of Kerry Group

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service