MilliporeSigma
All Photos(1)

686530

Supelco

Acetone solution

contains 20.0 % (v/v) acetonitrile, suitable for HPLC

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
MDL number:
PubChem Substance ID:

Quality Level

vapor density

2 (vs air)

vapor pressure

184 mmHg

form

liquid

contains

20.0 % (v/v) acetonitrile

expl. lim.

8 % (lit.)

technique(s)

HPLC: suitable

bp

56 °C/760 mmHg (lit.)

mp

-94 °C (lit.)

density

0.791 g/mL at 25 °C

application(s)

food and beverages

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is an acetone solution containing 20.0%v/v acetonitrile that can be employed for HPLC. Acetone is a water miscible organic solvent.

Packaging

4×4 L in glass bottle

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Abeer W Saeed et al.
Molecular pain, 11, 54-54 (2015-09-12)
Non-peptidergic nociceptive neurons are a sub-population of small diameter primary sensory neurons that comprise approximately 50 % of the C fiber population. Together with the peptidergic sub-population, they transmit nociceptive information from the periphery to the superficial dorsal horn of the
Gregory M Hayes et al.
American journal of translational research, 7(6), 1181-1188 (2015-08-19)
Soft tissue sarcoma (STS) is a heterogenous tumor arising from the embryonic mesoderm represented by approximately 50 histological subtypes. Effective therapeutic intervention is lacking for recurrent, late stage and metastatic disease. CD39, a cell-surface ectonucleotidase, has previously been shown to
Wenjie Zhang et al.
Oncotarget, 6(11), 9420-9433 (2015-04-01)
Autophagy plays a critical role in the progression of nonalcoholic steatohepatitis (NASH) and hepatocellular carcinoma (HCC). Protein tyrosine phosphatase receptor type O (PTPRO) was recently identified as a tumor suppressor, but little is known about its role in NASH. Here
Subhadip Ghatak et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 23(7), 1201-1210 (2015-04-22)
Tissue injury transiently silences miRNA-dependent posttranscriptional gene silencing in its effort to unleash adult tissue repair. Once the wound is closed, miRNA biogenesis is induced averting neoplasia. In this work, we report that Dicer plays an important role in reestablishing
Lukas von Tobel et al.
PLoS genetics, 10(11), e1004777-e1004777 (2014-11-21)
Centrioles are microtubule-based organelles important for the formation of cilia, flagella and centrosomes. Despite progress in understanding the underlying assembly mechanisms, how centriole integrity is ensured is incompletely understood, including in sperm cells, where such integrity is particularly critical. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service