MilliporeSigma
All Photos(1)

42146

Millipore

Agar

for membrane filtration, low gel strengh

Synonym(s):
Agar-agar, Gum agar
Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
NACRES:
NA.85

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

ign. residue

≤5.0%

loss

≤20% loss on drying

turbidity

≤15 NTU

pH

6.0-7.5 (1.5% in gel, after autoclaving)

mp

85-95 °C

transition temp

gel point 33-37 °C

gel strength

600-850 g/cm2

application(s)

microbiology

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service