All Photos(1)

17288

Supelco

Polyamide 6 on TLC-PET-foils

suitable for thin layer chromatography (TLC), PET foils

Synonym(s):
TLC-PET-foils Polyamide 6
NACRES:
NC.07

Quality Level

material

PET support

form

PET foils

feature

binder Proprietary
fluorescent indicator: no

application(s)

thin layer chromatography (TLC): suitable

L × W

20 cm × 20 cm

layer thickness

100 μm

particle size

15.0-25.0 μm

Application

Polyamide 6 on TLC-PET-foils may be used in TLC, LPLC, paper chromatography, PET foil backing.

Storage Class Code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service