MilliporeSigma
All Photos(1)

04669

Supelco

2,3,4,6-Tetra-O-(2-naphthoyl)-β-D-galactopyranosyl isothiocyanate

for chiral derivatization, ~90% (HPLC)

Empirical Formula (Hill Notation):
C51H35NO9S
CAS Number:
Molecular Weight:
837.89
MDL number:
PubChem Substance ID:

grade

for chiral derivatization

assay

~90% (HPLC)

SMILES string

O=C(OC[C@H]1O[C@@H](N=C=S)[C@H](OC(=O)c2ccc3ccccc3c2)[C@@H](OC(=O)c4ccc5ccccc5c4)[C@H]1OC(=O)c6ccc7ccccc7c6)c8ccc9ccccc9c8

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service