All Photos(1)

00765

Sigma-Aldrich

1-Ethyl-2,3-dimethylimidazolium trifluoromethanesulfonate

for electrochemistry, ≥99.0% (HPLC)

Empirical Formula (Hill Notation):
C8H13F3N2O3S
CAS Number:
Molecular Weight:
274.26
MDL number:
PubChem Substance ID:

grade

for electrochemistry

assay

≥99.0% (HPLC)

impurities

≤0.25% water

anion traces

bromide (Br-): ≤100 ppm
chloride (Cl-): ≤100 ppm
fluoride (F-): ≤100 ppm

SMILES string

[O-]S(=O)(=O)C(F)(F)F.CCn1cc[n+](C)c1C

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service