MilliporeSigma
All Photos(1)

11814150001

Roche

Anti-c-myc-Peroxidase

from mouse IgG1κ;

Synonym(s):
anti-myc-peroxidase

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9E10, monoclonal

Assay

90% (HPLC and SDS-PAGE)

form

solution

packaging

pkg of 500 μg (500 μl)

manufacturer/tradename

Roche

isotype

IgG1κ

epitope sequence

EQKLISEEDL

shipped in

dry ice

storage temp.

−20°C

General description

Anti-c-myc-Peroxidase is a monoclonal antibody to c-myc peptide (clone 9E10), conjugated to horseradish peroxidase. Anti-c-myc recognizes the 9E10 epitope (EQKLISEEDL), derived from the human c-myc protein. The monoclonal antibody against the c-myc epitope is well characterized and does not cross-react with other cellular proteins. The antibody recognizes its antigenic determinant even when the c-myc-peptide epitope is introduced into unrelated recombinant proteins by a technique known as "epitope tagging".

Specificity

Anti-c-myc-Peroxidase recognizes the 9E10 epitope (EQKLISEEDL), which was derived from the human c-myc protein. The monoclonal antibody against the c-myc epitope is well characterized and does not crossreact with other cellular proteins. The antibody recognizes its antigenic determinant even when the c-myc peptide epitope is introduced into unrelated recombinant proteins either N-terminal, C-terminal or internal.

Immunogen

Amino acid sequence EQKLISEEDL was derived from the human c-myc protein.

Application

  • Use Anti-c-myc-Peroxidase for the detection of native human c-myc protein and recombinant epitope-tagged proteins that contain the c-myc epitope using: Dot blots
  • Western blots

Quality

Function test: Western blot using a cell line that expresses a recombinant c-myc-tagged protein.

Preparation Note

Working concentration: Working concentration of conjugate depends on application and substrate.
The following concentrations should be taken as a guideline:
  • Western blot: 1 μg/ml

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(C)

No data available

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service