MilliporeSigma
All Photos(1)

PX1330

Supelco

Potassium Acetate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):
Potassium Acetate, Acetic Acid, Potassium Salt, Acetic Acid
Empirical Formula (Hill Notation):
C2H3KO2
CAS Number:
Molecular Weight:
98.14
MDL number:
EC Index Number:
204-822-2

Agency

meets USP/NF

Assay

≥99.0%

form

crystals

impurities

≤0.005% Insoluble matter

color

white

pH

7.5-8.5 (20 °C, 50 g/L in H2O)

mp

292 °C

solubility

2530 g/L

anion traces

chloride (Cl-): ≤0.003%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.005%
Fe: ≤5 ppm
Mg: ≤0.002%
Na: ≤0.03%
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

InChI

1S/C2H4O2.K/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

SCVFZCLFOSHCOH-UHFFFAOYSA-M

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay99.0% minCalcium (Ca)0.005% maxChloride (Cl)0.003% maxColorWhiteFormCrystals or crystalline powderHeavy Metals (by ICP-OES)5ppm maxInsoluble matter0.005% maxIron (Fe)5ppm maxMagnesium (Mg)0.002% maxPhosphate (PO4)0.001% maxSodium (Na)0.03% maxSulfate (SO4)0.002% maxpH of a 5% solution at 25°C6.5-9.0

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service