MilliporeSigma
All Photos(1)

FX0325

Sigma-Aldrich

Fluorescein, Sodium Salt

For the staining of cells, cell components, chromosomes, and bacteria

Synonym(s):
Fluorescein, Sodium Salt, Acid Yellow 73
Empirical Formula (Hill Notation):
C20H10Na2O5
CAS Number:
Molecular Weight:
376.27
MDL number:
EC Index Number:
208-253-0

Quality Level

form

powder

color

orange-red

pH

8.3 (20 °C, 10 g/L in H2O)

mp

>300 °C

solubility

500 g/L

density

1.601 g/cm3 at 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C20H12O5.2Na/c21-11-5-7-15-17(9-11)25-18-10-12(22)6-8-16(18)19(15)13-3-1-2-4-14(13)20(23)24;;/h1-10,21H,(H,23,24);;/q;2*+1/p-2

InChI key

NJDNXYGOVLYJHP-UHFFFAOYSA-L

General description

Fluorescein, Sodium Salt
FX0325-5 125g Poly Bottle

Packaging

125, 500 g in Plastic bottle

Application

For Professional Use.
For the staining of cells, cell components, chromosomes, and bacteria

Analysis Note

Assay40% minColorOrange-redFormPowderInfrared SpectrumConforms to standard

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

423.7 °F - Pensky-Martens closed cup

Flash Point(C)

217.6 °C - Pensky-Martens closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service