MilliporeSigma
All Photos(4)

EC222

Sigma-Aldrich

GenElute-E RNA Gel Loading Buffer

greener alternative

feature

Intended use (For stabilization of RNA samples for RNA gel electrophoresis)

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

greener alternative category

storage temp.

2-8°C

General description

For stabilization of RNA for RNA gel electrophoresis. The reagent comes ready-to-use as a 2x concentrated solution. The reagent contains glycerol for easy loading and bromo-phenol blue for visualization of the electrophoresis migration. Per 500 μL of reagent, 100 RNA samples can be analyzed (if used at 10 μL per well).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is aligned with "Waste prevention" and "Designing safer chemicals". Click here for more information.

Storage and Stability

Use within 12 months of receipt.

Other Notes

We believe that green chemistry will contribute to a better tomorrow. With a growing portfolio of greener alternatives, there are now more choices to reduce the ecological impact of your research while still delivering quality and efficacy so your results are not compromised. GenElute-E kits adhere to the principles of SMASH Packaging, our global initiative to improve the sustainability through the reduction of packaging materials, the increased use of sustainable alternatives, and improved recyclability. Packaging is FCS-marked for sustainable forestry practices and produced from greater than 70% recycled content.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Product Information Sheet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service