MilliporeSigma
All Photos(1)

BX0360

Supelco

Benzoic Acid

Meets ACS Specifications GR ACS

Synonym(s):
Benzoic Acid
Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
EC Index Number:
200-618-2

vapor pressure

0.001 hPa ( 20 °C)

Quality Level

assay

≥99.5%

form

crystalline powder

impurities

≤0.002% Sulfur compounds (as S)
≤0.005% Insoluble in methanol

ign. residue

≤0.005%

color

white

pH

2.5-3.5 (20 °C in H2O, saturated solution)

bp

249 °C/1013 hPa

mp

121-123 °C

transition temp

flash point 121 °C

solubility

2.9 g/L

density

1.26 g/cm3 at 15 °C

anion traces

chloride (Cl-): ≤0.005%

cation traces

Fe: ≤2 ppm
heavy metals (as Pb): ≤5 ppm

shipped in

ambient

storage temp.

room temp

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Packaging

500 g in Plastic bottle

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay99.5% minChlorine compounds (as Cl)0.005% maxColorWhiteFormCrystalline powderFreezing point122-123°CHeavy metals (as Pb)5ppm maxIdentity (IR-spectrum)ConformsInsoluble in methanol0.005% maxIron (Fe)2ppm maxResidue after ignition0.005% maxSubstances reducing permanganateTo pass testSulfur compounds (as S)0.002% max

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service