MilliporeSigma
All Photos(1)

BM-113010NL

Millipore

Auto2D® IEF Chip pH3-10NL

new

precast gel, used for first-dimension separations on the Auto2D® 2-D Electrophoresis Device.

Synonym(s):
2D Electrophoresis, 2DE, IEF, IEF Chip, IPG gel, precast gel

product line

Auto2D®

General description

IEF Chip is 1st dimensional gel (separation medium) composed of IPG strip gel. IEF Chip pH3-10 is used for first-dimension separations on the Auto2D® 2-D Electrophoresis Device. This gives comprehensive view of whole pH range with expansion of acidic region.

Application

  • Differential proteomic analysis
  • Post-translational modification
  • Protein Isoforms
  • Western Blotting

Legal Information

Auto2D is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service