MilliporeSigma
All Photos(1)

A-124

Supelco

Multi-component Alcohol Mix

250 μg/mL each component in H2O, certified reference material, Cerilliant®

NACRES:
NA.24

Quality Level

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Acetone
Ethanol
Isopropanol
Methanol

packaging

ampule of 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

250 μg/mL each component in H2O

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

multi-component solution

storage temp.

2-8°C

InChI

1S/C15H13N3O/c16-11-6-7-13-12(8-11)15(17-9-14(19)18-13)10-4-2-1-3-5-10/h1-8H,9,16H2,(H,18,19)/i1D,2D,3D,4D,5D

InChI key

OYOUQHVDCKOOAL-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A multicomponent calibration kit containing acetone, ethanol, isopropanol, and methanol at a concentration of 250 μg/mL. This kit is suitable for calibration of blood alcohol testing methods using GC Headspace.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Cerilliant Corporation

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service