NCXAGPD30

Sigma-Aldrich

Silver Nanospheres

30 nm, NanoXact, Dried, PVP 40 kDa (Polymer), powder

NACRES:
NA.23

form

nanospheres
powder

packaging

pkg of SKU size by silver mass

particle size

30 nm

Looking for similar products? Visit Product Comparison Guide

Application

Owing to their plasmonic properties nanoparticles of gold and silver find broad range of applications extending from diagnostics to molecular detection, photothermal and microwave-based therapies, and nanotoxicology. Colloidal silver nanoparticles also find applications in antimicrobial coatings, electronics and photovoltaics, and surface enhanced Raman spectroscopy (SERS) whereas the colloidal gold nanoparticles find applications in lateral flow assays (immunochromatographic assays) and as TEM labels and size standards.

Our NanoXact materials are precisely engineered, unagglomerated silver nanospheres with a narrow size distributions (CVs <15% for 30-200 nm sizes, <20% for 20 nm, and <25% for 5 and 10 nm sizes). These nanospheres contain PVP (40 kD polyvinylpyrrolidone) as excipient. The PVP enhances stability and dispersibility of NanpXact silver nanospheres. These are redispersible in a variety of polar solvents such as water, alcohols, DMF, and DMSO. They show medium dispersibility in chloroform, and low dispersibility in dichloromethane.

Storage and Stability

DO NOT FREEZE. Store at 2-25C in a dry place. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Legal Information

Product of nanoComposix, Inc.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

hazcat

Aquatic Acute 1 - Aquatic Chronic 1

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service