MilliporeSigma
All Photos(2)

GF48834311

Silver

insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyimide insulation, 99.9%

Synonym(s):
Silver, AG005842
Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
PubChem Substance ID:
NACRES:
NA.23

assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 488-343-11

resistivity

1.59 μΩ-cm, 20°C

L × diam. × thickness

10 m × 0.125 mm × 0.0125 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service