MilliporeSigma
All Photos(1)

612596

Supelco

Nicotine solution

NMR reference standard, 20 mg/mL in ethanol-d6 (99.5 atom % D), TMS 0.05 %, NMR tube size 5 mm × 8 in.

Empirical Formula (Hill Notation):
C10H14N2
Molecular Weight:
162.23
NACRES:
NA.24

grade

NMR reference standard

Quality Level

composition

TMS, 0.05%

concentration

20 mg/mL in ethanol-d6 (99.5 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

format

single component solution

Looking for similar products? Visit Product Comparison Guide

Related Categories

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service