MilliporeSigma
All Photos(1)

270520

Supelco

Ethyl acetate

HPLC grade, 99.8%

Synonym(s):
EtOAc
Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
PubChem Substance ID:

grade

HPLC grade

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

assay

99.8%

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

HPLC: suitable

impurities

<0.050% water

evapn. residue

<0.0003%

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)
76.5-77.5 °C

mp

−84 °C (lit.)

solubility

H2O: soluble

density

0.902 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 254 nm Amax: 1.00
λ: 263 nm Amax: 0.05
λ: 275-400 nm Amax: 0.01

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service